Product Details

SNP ID
rs56675391
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:93152199 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAAATGCATATCAGAGCAAACTC[C/T]AGGCCTTAGTGTGAGGTGAACACGG
Phenotype
MIM: 616876
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMED5 PubMed Links

Gene Details

Gene
TMED5
Gene Name
transmembrane p24 trafficking protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167830.1 3657 UTR 3 NP_001161302.1
NM_016040.4 3657 UTR 3 NP_057124.3

View Full Product Details