Product Details

SNP ID
rs61753533
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:111296460 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGAGGTTAGCACTCCAGCCTGGG[C/T]GACTGAGCAAGACTCCATCTCAAAA
Phenotype
MIM: 615693 MIM: 615694
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COLCA1 PubMed Links

Gene Details

Gene
COLCA1
Gene Name
colorectal cancer associated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302644.1 2428 Missense Mutation ACC,GCC T7A NP_001289573.1
NM_001302645.1 2428 Missense Mutation ACC,GCC T7A NP_001289574.1
NM_001302646.1 2428 Missense Mutation ACC,GCC T7A NP_001289575.1
NM_001302647.1 2428 Missense Mutation ACC,GCC T7A NP_001289576.1
NM_001302648.1 2428 Missense Mutation ACC,GCC T7A NP_001289577.1
NM_207429.3 2428 Missense Mutation ACC,GCC T7A NP_997312.1
Gene
COLCA2
Gene Name
colorectal cancer associated 2
There are no transcripts associated with this gene.

View Full Product Details