Product Details
- SNP ID
-
rs61910591
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:124611635 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGACCGCAGGGGACCCCGCCTCAAA[A/G]GACTGCGTCTGGAACTGCCCCTGGA
- Phenotype
-
MIM: 608422
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PANX3
PubMed Links
Gene Details
- Gene
- PANX3
- Gene Name
- pannexin 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_052959.2 |
79 |
Missense Mutation |
AGA,GGA |
R27G |
NP_443191.1 |
View Full Product Details