Product Details

SNP ID
hCV90489850
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:105041675 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAGCTTGATTCTGCCTTCTGGGC[C/T]TGAGCATGTGGGCATAGCTGGGTTG
Phenotype
MIM: 615680 MIM: 147678
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CARD16 PubMed Links

Gene Details

Gene
CARD16
Gene Name
caspase recruitment domain family member 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017534.1 409 Missense Mutation AGC,GGC S107G NP_001017534.1
NM_052889.2 409 UTR 3 NP_443121.1
Gene
CASP1
Gene Name
caspase 1
There are no transcripts associated with this gene.

View Full Product Details