Product Details

SNP ID
rs61284324
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:85958908 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCAATTTCCTTCATGGGCCTTCA[C/T]TGAGTTACTTGTAGCATTCTAATGG
Phenotype
MIM: 603025
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PICALM PubMed Links

Gene Details

Gene
PICALM
Gene Name
phosphatidylinositol binding clathrin assembly protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008660.2 2386 UTR 3 NP_001008660.1
NM_001206946.1 2386 UTR 3 NP_001193875.1
NM_001206947.1 2386 UTR 3 NP_001193876.1
NM_007166.3 2386 UTR 3 NP_009097.2
XM_005274322.2 2386 UTR 3 XP_005274379.1
XM_005274323.2 2386 UTR 3 XP_005274380.1
XM_005274324.2 2386 UTR 3 XP_005274381.1
XM_005274325.2 2386 UTR 3 XP_005274382.1
XM_005274326.2 2386 UTR 3 XP_005274383.1
XM_005274327.2 2386 UTR 3 XP_005274384.1
XM_005274328.2 2386 UTR 3 XP_005274385.1
XM_005274329.3 2386 UTR 3 XP_005274386.1
XM_005274330.2 2386 UTR 3 XP_005274387.1
XM_005274331.2 2386 UTR 3 XP_005274388.1
XM_005274332.2 2386 UTR 3 XP_005274389.1
XM_005274333.2 2386 UTR 3 XP_005274390.1
XM_005274334.2 2386 UTR 3 XP_005274391.1
XM_005274335.2 2386 UTR 3 XP_005274392.1
XM_005274336.2 2386 UTR 3 XP_005274393.1
XM_005274337.2 2386 UTR 3 XP_005274394.1
XM_005274340.2 2386 UTR 3 XP_005274397.1
XM_006718699.2 2386 UTR 3 XP_006718762.1
XM_006718700.2 2386 UTR 3 XP_006718763.1
XM_006718701.3 2386 UTR 3 XP_006718764.1
XM_011545293.1 2386 UTR 3 XP_011543595.1
XM_017018381.1 2386 UTR 3 XP_016873870.1
XM_017018382.1 2386 UTR 3 XP_016873871.1
XM_017018383.1 2386 UTR 3 XP_016873872.1
XM_017018384.1 2386 UTR 3 XP_016873873.1
XM_017018385.1 2386 UTR 3 XP_016873874.1
XM_017018386.1 2386 UTR 3 XP_016873875.1
XM_017018387.1 2386 UTR 3 XP_016873876.1
XM_017018388.1 2386 UTR 3 XP_016873877.1

View Full Product Details