Product Details

SNP ID
rs61115239
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:75155919 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAGGGAATAATGCTCACTAGGGT[A/G]GGGGGTACGTGGGATGGGACTGGCT
Phenotype
MIM: 604988
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLCO2B1 PubMed Links

Gene Details

Gene
SLCO2B1
Gene Name
solute carrier organic anion transporter family member 2B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145211.2 Intron NP_001138683.1
NM_001145212.2 Intron NP_001138684.1
NM_007256.4 Intron NP_009187.1
XM_017017157.1 Intron XP_016872646.1

View Full Product Details