Product Details

SNP ID
rs58005372
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:132563310 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAAGAGATAAGATCCATTGCCCA[A/G]CTGGAGGGGATACCAGGAACATCCA
Phenotype
MIM: 147850 MIM: 604040
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IL5 PubMed Links

Gene Details

Gene
IL5
Gene Name
interleukin 5
There are no transcripts associated with this gene.

Gene
RAD50
Gene Name
RAD50 double strand break repair protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005732.3 Intron NP_005723.2

View Full Product Details