Product Details

SNP ID
rs61761877
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:2833917 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTGAGAGTGTAACTCTCTTCCAG[A/T]TTGAACCTGGGCAAGCTGACATTAA
Phenotype
MIM: 130135
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
SERPINB1 PubMed Links

Gene Details

Gene
SERPINB1
Gene Name
serpin family B member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030666.3 1211 Missense Mutation AAA,AAT K277N NP_109591.1
XM_011514333.1 1211 Missense Mutation AAA,AAT K277N XP_011512635.1
XM_011514334.2 1211 Missense Mutation AAA,AAT K277N XP_011512636.1
XM_011514335.2 1211 Missense Mutation AAA,AAT K228N XP_011512637.1

View Full Product Details