Product Details

SNP ID
rs72710522
Assay Type
Functionally tested
NCBI dbSNP Submissions
28
Location
Chr.1:170936242 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAAGTCCTGGATTCTGCTTTCTG[C/T]AAGACAGAGACCCAGGAGATCCAAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MROH9 PubMed Links

Gene Details

Gene
MROH9
Gene Name
maestro heat like repeat family member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163629.1 Intron NP_001157101.1
NM_025063.2 Intron NP_079339.2
XM_011510005.2 Intron XP_011508307.1
XM_011510006.2 Intron XP_011508308.1
XM_011510007.2 Intron XP_011508309.1

View Full Product Details