Product Details

SNP ID
rs72549320
Assay Type
Functionally Tested
NCBI dbSNP Submissions
4
Location
Chr.1:171092752 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGGGGCTGGAGCCCACCTGCTTT[A/G]AGAAGAGCAATGACATTGGGGGCCT
Phenotype
MIM: 136132
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
FMO3 PubMed Links

Gene Details

Gene
FMO3
Gene Name
flavin containing monooxygenase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002294.2 205 Missense Mutation AAG,GAG K32E NP_001002294.1
NM_001319173.1 205 UTR 5 NP_001306102.1
NM_001319174.1 205 Missense Mutation AAG,GAG K32E NP_001306103.1
NM_006894.5 205 Missense Mutation AAG,GAG K32E NP_008825.4
XM_011509345.2 205 UTR 5 XP_011507647.1
Gene
MIR1295A
Gene Name
microRNA 1295a
There are no transcripts associated with this gene.

Gene
MIR1295B
Gene Name
microRNA 1295b
There are no transcripts associated with this gene.

View Full Product Details