Product Details

SNP ID
rs73075725
Assay Type
Functionally tested
NCBI dbSNP Submissions
30
Location
Chr.1:203167962 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCATTCCTGAGTTCTGGACACCAG[C/T]CTCTGGGGAAGGGAGGCCAGCAGGT
Phenotype
MIM: 102775 MIM: 160795
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADORA1 PubMed Links

Gene Details

Gene
ADORA1
Gene Name
adenosine A1 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000674.2 1655 Intron NP_000665.1
NM_001048230.1 1655 Intron NP_001041695.1
XM_005244901.1 1655 Intron XP_005244958.1
XM_005244902.4 1655 Intron XP_005244959.1
XM_017000342.1 1655 Intron XP_016855831.1
Gene
MYBPH
Gene Name
myosin binding protein H
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004997.2 1655 UTR 3 NP_004988.2

View Full Product Details