Product Details

SNP ID
rs73140586
Assay Type
Functionally tested
NCBI dbSNP Submissions
32
Location
Chr.1:247451355 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAGAGCCAGGGGCACCAACACGA[A/G]GAAGGCCACCAGCACAGCCAGTATG
Phenotype
MIM: 606416
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NLRP3 PubMed Links

Gene Details

Gene
NLRP3
Gene Name
NLR family pyrin domain containing 3
There are no transcripts associated with this gene.

Gene
OR2B11
Gene Name
olfactory receptor family 2 subfamily B member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004492.1 628 Missense Mutation CTC,TTC L210F NP_001004492.1

View Full Product Details