Product Details
- SNP ID
-
rs74153215
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
31
- Location
-
Chr.1:247965423 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTGGGACAGATTTTCTACTTGTTG[A/G]TCTTTTCCAATATGGCTGGATAAAC
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
OR2AK2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs41304159] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR2AK2
- Gene Name
- olfactory receptor family 2 subfamily AK member 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001004491.1 |
92 |
Missense Mutation |
GAT,GGT |
D31G |
NP_001004491.1 |
- Gene
- OR2L13
- Gene Name
- olfactory receptor family 2 subfamily L member 13
View Full Product Details