Product Details

SNP ID
rs71424910
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:236332275 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAGATATTTCAATCTGAACACAG[A/G]GATGCTGTGAAAATGAATTATGAAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IQCA1 PubMed Links

Gene Details

Gene
IQCA1
Gene Name
IQ motif containing with AAA domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270584.1 Intron NP_001257513.1
NM_001270585.1 Intron NP_001257514.1
NM_024726.4 Intron NP_079002.3
XM_011511864.1 Intron XP_011510166.1
XM_011511865.2 Intron XP_011510167.1
XM_011511866.1 Intron XP_011510168.1
XM_017004960.1 Intron XP_016860449.1

View Full Product Details