Product Details

SNP ID
rs73978120
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:175927485 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGTTTGGGAAGGCATCTCTCTGA[C/G]GAGGCAAAATTTAAGCTGAAATCTG
Phenotype
MIM: 610236
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LNPK PubMed Links

Gene Details

Gene
LNPK
Gene Name
lunapark, ER junction formation factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001305008.1 4110 UTR 3 NP_001291937.1
NM_001305009.1 4110 UTR 3 NP_001291938.1
NM_001305010.1 4110 UTR 3 NP_001291939.1
NM_001305011.1 4110 UTR 3 NP_001291940.1
NM_030650.2 4110 UTR 3 NP_085153.1
XM_005246878.3 4110 UTR 3 XP_005246935.1
XM_005246879.4 4110 UTR 3 XP_005246936.1
XM_005246880.3 4110 UTR 3 XP_005246937.1
XM_006712783.2 4110 UTR 3 XP_006712846.1
XM_011511945.2 4110 UTR 3 XP_011510247.1
XM_017005055.1 4110 UTR 3 XP_016860544.1

View Full Product Details