Product Details

SNP ID
rs72922378
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:199761696 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTGTAAGTAGGGTAAAATCCCAA[A/G]CCCCAACATCTTCATCCACTACACC
Phenotype
MIM: 614308
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FTCDNL1 PubMed Links

Gene Details

Gene
FTCDNL1
Gene Name
formiminotransferase cyclodeaminase N-terminal like
There are no transcripts associated with this gene.

View Full Product Details