Product Details

SNP ID
rs72548794
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:223052254 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGACGATTTGGGAATTCAAAACT[C/T]GGGACAAACTGTCAGCCTTGGTAAG
Phenotype
MIM: 607775
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCNE4 PubMed Links

Gene Details

Gene
KCNE4
Gene Name
potassium voltage-gated channel subfamily E regulatory subunit 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080671.3 325 Missense Mutation TCG,TTG S37L NP_542402.3

View Full Product Details