Product Details
- SNP ID
-
rs72548795
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:223053061 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCTGCTGCTGTACAAAGACGAGGA[A/G]CGGCTCTGGGGGGAGGCCATGAAGC
- Phenotype
-
MIM: 607775
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
KCNE4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs10201907] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KCNE4
- Gene Name
- potassium voltage-gated channel subfamily E regulatory subunit 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_080671.3 |
599 |
Silent Mutation |
GAA,GAG |
E128E |
NP_542402.3 |
View Full Product Details