Product Details
- SNP ID
-
rs73216700
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:109146351 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAGCTATTTACCTTGACATCAGTAA[A/G]TGCGTATAATGCATTCTATAAGTTA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FLJ22763
PubMed Links
Gene Details
- Gene
- FLJ22763
- Gene Name
- uncharacterized LOC401081
There are no transcripts associated with this gene.
View Full Product Details