Product Details

SNP ID
rs72655938
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:119212639 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTATAAGAACAACTACTCAACCC[A/G]TCTGTTCTCCAGACTCGTGACACTT
Phenotype
MIM: 604015 MIM: 602380
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
B4GALT4 PubMed Links

Gene Details

Gene
B4GALT4
Gene Name
beta-1,4-galactosyltransferase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003778.3 1364 Silent Mutation GAC,GAT D315D NP_003769.1
NM_212543.1 1364 Silent Mutation GAC,GAT D315D NP_997708.1
XM_005247855.1 1364 Silent Mutation GAC,GAT D315D XP_005247912.1
XM_006713798.2 1364 Silent Mutation GAC,GAT D315D XP_006713861.1
XM_006713799.2 1364 Silent Mutation GAC,GAT D315D XP_006713862.1
XM_006713800.1 1364 Silent Mutation GAC,GAT D315D XP_006713863.1
XM_006713801.2 1364 Silent Mutation GAC,GAT D315D XP_006713864.1
XM_011513260.1 1364 Silent Mutation GAC,GAT D315D XP_011511562.1
XM_017007401.1 1364 Silent Mutation GAC,GAT D274D XP_016862890.1
Gene
UPK1B
Gene Name
uroplakin 1B
There are no transcripts associated with this gene.

View Full Product Details