Product Details

SNP ID
rs73176886
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:168252862 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATTCTGTGGTGGGTATGGAGAGA[G/T]GTTGGCATGCTAATTGTGCTGCTAT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
EGFEM1P PubMed Links

Gene Details

Gene
EGFEM1P
Gene Name
EGF like and EMI domain containing 1, pseudogene
There are no transcripts associated with this gene.

View Full Product Details