Product Details

SNP ID
rs73884599
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:183715156 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTGCTTCACAGTGAAGAACTGAA[G/T]GTCATCATGTCTGGAATCAAGCGAA
Phenotype
MIM: 613373
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
YEATS2 PubMed Links

Gene Details

Gene
YEATS2
Gene Name
YEATS domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018023.4 100 UTR 5 NP_060493.3
XM_005247597.2 100 UTR 5 XP_005247654.1
XM_005247598.4 100 UTR 5 XP_005247655.1
XM_006713688.3 100 UTR 5 XP_006713751.1
XM_011512965.1 100 UTR 5 XP_011511267.1
XM_011512966.1 100 UTR 5 XP_011511268.1
XM_017006809.1 100 UTR 5 XP_016862298.1
XM_017006810.1 100 UTR 5 XP_016862299.1
XM_017006811.1 100 Intron XP_016862300.1
XM_017006812.1 100 UTR 5 XP_016862301.1
XM_017006813.1 100 UTR 5 XP_016862302.1
XM_017006814.1 100 UTR 5 XP_016862303.1

View Full Product Details