Product Details

SNP ID
rs73057404
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:186550900 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCCCAGGCAAAAAGCAAAATAAG[A/G]CAAAATTTGGCCATTATATTGGCCC
Phenotype
MIM: 123730
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CRYGS PubMed Links

Gene Details

Gene
CRYGS
Gene Name
crystallin gamma S
There are no transcripts associated with this gene.

Gene
TBCCD1
Gene Name
TBCC domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134415.1 Intron NP_001127887.1
NM_001286749.1 Intron NP_001273678.1
NM_018138.4 Intron NP_060608.1

View Full Product Details