Product Details

SNP ID
rs73889748
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:189601570 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCTGCAGAAAGAATCCTCATTTT[A/C]TTGGATTACTATTTTTATCCTTAGA
Phenotype
MIM: 603273
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
TP63 PubMed Links
Additional Information
For this assay, SNP(s) [rs11354146] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
TP63
Gene Name
tumor protein p63
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114978.1 Intron NP_001108450.1
NM_001114979.1 Intron NP_001108451.1
NM_001114980.1 Intron NP_001108452.1
NM_001114981.1 Intron NP_001108453.1
NM_001114982.1 Intron NP_001108454.1
NM_003722.4 Intron NP_003713.3
XM_005247843.3 Intron XP_005247900.1
XM_005247844.4 Intron XP_005247901.1
XM_011513251.2 Intron XP_011511553.1
XM_011513252.2 Intron XP_011511554.1
XM_017007387.1 Intron XP_016862876.1

View Full Product Details