Product Details

SNP ID
rs73196068
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:6028962 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTTGAATGAAGAGATGCACACCA[C/T]GTGGTCCCTGTCCTCAAGGACTCAC
Phenotype
MIM: 611195
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
JAKMIP1 PubMed Links

Gene Details

Gene
JAKMIP1
Gene Name
janus kinase and microtubule interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099433.1 Intron NP_001092903.1
NM_001306133.1 Intron NP_001293062.1
NM_001306134.1 Intron NP_001293063.1
NM_144720.3 Intron NP_653321.1
XM_011513400.2 Intron XP_011511702.1
XM_017007788.1 Intron XP_016863277.1
XM_017007789.1 Intron XP_016863278.1
XM_017007790.1 Intron XP_016863279.1
XM_017007791.1 Intron XP_016863280.1
XM_017007792.1 Intron XP_016863281.1
XM_017007793.1 Intron XP_016863282.1
XM_017007794.1 Intron XP_016863283.1
XM_017007795.1 Intron XP_016863284.1
XM_017007796.1 Intron XP_016863285.1
XM_017007797.1 Intron XP_016863286.1
XM_017007798.1 Intron XP_016863287.1
XM_017007799.1 Intron XP_016863288.1

View Full Product Details