Product Details

SNP ID
rs73824590
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:73986864 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTAAATATATTAACAAAAATGT[A/G]TTGAGCAAATATTCTCTGTTGGGCT
Phenotype
MIM: 600324 MIM: 173460 MIM: 121010
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CXCL5 PubMed Links

Gene Details

Gene
CXCL5
Gene Name
C-X-C motif chemokine ligand 5
There are no transcripts associated with this gene.

Gene
PF4
Gene Name
platelet factor 4
There are no transcripts associated with this gene.

Gene
PPBP
Gene Name
pro-platelet basic protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002704.3 882 UTR 3 NP_002695.1

View Full Product Details