Product Details

SNP ID
rs73235697
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:40815416 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTTTTGTTCGATTTTAATTCCTC[A/G]AATTTAGTTTTTCATCAATTTTGAA
Phenotype
MIM: 602710
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
APBB2 PubMed Links

Gene Details

Gene
APBB2
Gene Name
amyloid beta precursor protein binding family B member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166050.1 3526 UTR 3 NP_001159522.1
NM_001166051.1 3526 UTR 3 NP_001159523.1
NM_001166052.1 3526 UTR 3 NP_001159524.1
NM_001166053.1 3526 UTR 3 NP_001159525.1
NM_001166054.1 3526 UTR 3 NP_001159526.1
NM_004307.1 3526 UTR 3 NP_004298.1
NM_173075.4 3526 UTR 3 NP_775098.2
XM_005248101.3 3526 UTR 3 XP_005248158.1
XM_006714005.3 3526 UTR 3 XP_006714068.1
XM_006714007.3 3526 UTR 3 XP_006714070.1
XM_006714008.3 3526 UTR 3 XP_006714071.1
XM_006714009.3 3526 UTR 3 XP_006714072.1
XM_006714010.3 3526 UTR 3 XP_006714073.1
XM_006714011.3 3526 UTR 3 XP_006714074.1
XM_006714012.3 3526 UTR 3 XP_006714075.1
XM_011513686.2 3526 UTR 3 XP_011511988.1
XM_011513687.2 3526 UTR 3 XP_011511989.1
XM_011513688.2 3526 Intron XP_011511990.1
XM_017008142.1 3526 UTR 3 XP_016863631.1
XM_017008143.1 3526 UTR 3 XP_016863632.1
XM_017008144.1 3526 UTR 3 XP_016863633.1
XM_017008145.1 3526 UTR 3 XP_016863634.1
XM_017008146.1 3526 UTR 3 XP_016863635.1
XM_017008147.1 3526 UTR 3 XP_016863636.1
XM_017008148.1 3526 UTR 3 XP_016863637.1
XM_017008149.1 3526 UTR 3 XP_016863638.1
XM_017008150.1 3526 UTR 3 XP_016863639.1
XM_017008151.1 3526 UTR 3 XP_016863640.1
XM_017008152.1 3526 UTR 3 XP_016863641.1
XM_017008153.1 3526 UTR 3 XP_016863642.1
XM_017008154.1 3526 UTR 3 XP_016863643.1
XM_017008155.1 3526 UTR 3 XP_016863644.1
XM_017008156.1 3526 UTR 3 XP_016863645.1
XM_017008157.1 3526 UTR 3 XP_016863646.1
XM_017008158.1 3526 UTR 3 XP_016863647.1
XM_017008159.1 3526 Intron XP_016863648.1
Gene
NSUN7
Gene Name
NOP2/Sun RNA methyltransferase family member 7
There are no transcripts associated with this gene.

View Full Product Details