Product Details
- SNP ID
-
rs73736824
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:6310851 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAATCAAGGTAAAATCAGGCCTAAT[A/G]GCCTCAGGGTTTGGGCCCTAACCAA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FLJ33360
PubMed Links
Gene Details
- Gene
- FLJ33360
- Gene Name
- FLJ33360 protein
There are no transcripts associated with this gene.
View Full Product Details