Product Details

SNP ID
rs73492614
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:89928294 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCCTAGTTTAAATGGGGCATAAT[A/G]TAGAGAAGTAGGTAGCCAAATAGGG
Phenotype
MIM: 605394
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BACH2 PubMed Links

Gene Details

Gene
BACH2
Gene Name
BTB domain and CNC homolog 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170794.1 8486 UTR 3 NP_001164265.1
NM_021813.3 8486 UTR 3 NP_068585.1
XM_005248758.4 8486 UTR 3 XP_005248815.1
XM_005248759.4 8486 UTR 3 XP_005248816.1
XM_011536039.2 8486 Intron XP_011534341.1
XM_011536040.2 8486 Intron XP_011534342.1
XM_017011156.1 8486 UTR 3 XP_016866645.1
XM_017011157.1 8486 UTR 3 XP_016866646.1
XM_017011158.1 8486 UTR 3 XP_016866647.1
XM_017011159.1 8486 UTR 3 XP_016866648.1
XM_017011160.1 8486 UTR 3 XP_016866649.1
XM_017011161.1 8486 UTR 3 XP_016866650.1
XM_017011162.1 8486 UTR 3 XP_016866651.1
XM_017011163.1 8486 UTR 3 XP_016866652.1
XM_017011164.1 8486 UTR 3 XP_016866653.1
XM_017011165.1 8486 Intron XP_016866654.1
XM_017011166.1 8486 UTR 3 XP_016866655.1
XM_017011167.1 8486 Intron XP_016866656.1

View Full Product Details