Product Details

SNP ID
rs73745010
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:34589013 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCTGCCTGTCTCACACCCTCAC[C/T]CACCACCCAGCTGGATGGGGGTGTG
Phenotype
MIM: 612217
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C6orf106 PubMed Links

Gene Details

Gene
C6orf106
Gene Name
chromosome 6 open reading frame 106
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022758.5 2498 UTR 3 NP_073595.2
NM_024294.3 2498 UTR 3 NP_077270.1
XM_005249298.2 2498 Intron XP_005249355.1

View Full Product Details