Product Details

SNP ID
rs72840230
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:16304091 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGTTTTTGGTACACCCCAGAAAA[C/T]GGTTTATAAAAATCATTAGTCCTGG
Phenotype
MIM: 601556 MIM: 139265
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATXN1 PubMed Links

Gene Details

Gene
ATXN1
Gene Name
ataxin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000332.3 5657 UTR 3 NP_000323.2
NM_001128164.1 5657 UTR 3 NP_001121636.1
Gene
GMPR
Gene Name
guanosine monophosphate reductase
There are no transcripts associated with this gene.

View Full Product Details