Product Details

SNP ID
rs73687837
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:30371546 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGACCACAGCAGATGGAGCTGGAG[G/T]AGGAAAAGGCAGGCAGTGGACTCTG
Phenotype
MIM: 612061
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DKFZP586I1420 PubMed Links

Gene Details

Gene
DKFZP586I1420
Gene Name
uncharacterized protein DKFZp586I1420
There are no transcripts associated with this gene.

Gene
ZNRF2
Gene Name
zinc and ring finger 2, E3 ubiquitin protein ligase
There are no transcripts associated with this gene.

View Full Product Details