Product Details

SNP ID
rs73336454
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:47659677 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCCTTCCTGGAGGCCAGGATGCC[A/G]AGAACTGCCTCCTAGCCACCCGCTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C7orf65 PubMed Links

Gene Details

Gene
C7orf65
Gene Name
chromosome 7 open reading frame 65
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001123065.1 940 UTR 3 NP_001116537.1

View Full Product Details