Product Details
- SNP ID
-
rs71563577
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:98619844 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTGAACGCGCTGCTGCAGAGGGTC[A/C]CCGAGCTGGAGCGAGGTGTGTGCCT
- Phenotype
-
MIM: 600750
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
NPTX2
PubMed Links
Gene Details
- Gene
- NPTX2
- Gene Name
- neuronal pentraxin 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002523.2 |
805 |
Missense Mutation |
ACC,CCC |
T210P |
NP_002514.1 |
View Full Product Details