Product Details

SNP ID
rs73720339
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:124031764 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGTAAAGCGTATAAAAATTAAAGG[A/G]CTAAAAAACCCACTTCATTTTAAAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC105375483 PubMed Links

Gene Details

Gene
LOC105375483
Gene Name
uncharacterized LOC105375483
There are no transcripts associated with this gene.

Gene
TMEM229A
Gene Name
transmembrane protein 229A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136002.1 1706 UTR 3 NP_001129474.1

View Full Product Details