Product Details

SNP ID
rs73255219
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:86048549 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGATTTGTGAGAATAATGTGACTT[A/G]GAAGACTGTGCAGATCCAGGACTCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PSKH2 PubMed Links

Gene Details

Gene
PSKH2
Gene Name
protein serine kinase H2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033126.1 1071 Silent Mutation TCC,TCT S357S NP_149117.1
XM_017013929.1 1071 Silent Mutation TCC,TCT S476S XP_016869418.1

View Full Product Details