Product Details
- SNP ID
-
rs73692313
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:90623861 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGTTGTCAGGTTAGACAGAAAAAA[A/T]TAAATCAAATGTATTGTTGGTTGGG
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM64
PubMed Links
Gene Details
- Gene
- TMEM64
- Gene Name
- transmembrane protein 64
View Full Product Details