Product Details
- SNP ID
-
rs72631813
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:25678198 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGCCCACGCCGCCGCCGCCGCCGC[C/T]GGGGCTGCCCCCACCACGAGCCCGG
- Phenotype
-
MIM: 610529
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TUSC1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs72631814] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TUSC1
- Gene Name
- tumor suppressor candidate 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001004125.2 |
661 |
Missense Mutation |
AGC,GGC |
S42G |
NP_001004125.1 |
View Full Product Details