Product Details

SNP ID
rs73663505
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:104694560 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGCCATGACTGAATTCTTTCTG[A/G]TGGGGCTTTCCCAATATCCAGAGCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR13D1 PubMed Links

Gene Details

Gene
OR13D1
Gene Name
olfactory receptor family 13 subfamily D member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004484.1 139 Missense Mutation ATG,GTG M47V NP_001004484.1

View Full Product Details