Product Details
- SNP ID
-
rs74131326
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:29294088 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGGTAGAGATAGAGAAAAGTTATC[A/C]CAGGCCAGGGGAAGAGCAAAAGCAA
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
LYZL1
PubMed Links
Gene Details
- Gene
- LYZL1
- Gene Name
- lysozyme like 1
View Full Product Details