Product Details

SNP ID
rs73426354
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:3677879 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTACTAGGAGTTGGAACTTTGATGA[A/G]GATAATAATAGATAACAATTATTAA
Phenotype
MIM: 606372 MIM: 601021
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHRNA10 PubMed Links

Gene Details

Gene
CHRNA10
Gene Name
cholinergic receptor nicotinic alpha 10 subunit
There are no transcripts associated with this gene.

Gene
NUP98
Gene Name
nucleoporin 98
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005387.6 Intron NP_005378.4
NM_016320.4 Intron NP_057404.2
NM_139131.4 Intron NP_624357.1
NM_139132.3 Intron NP_624358.2

View Full Product Details