Product Details

SNP ID
rs72484720
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:7796118 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGGTTCAACATGGGAATCACCAC[C/T]GTGTACAACACAGACACCACCTTGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR5P2 PubMed Links

Gene Details

Gene
OR5P2
Gene Name
olfactory receptor family 5 subfamily P member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153444.1 825 Silent Mutation ACA,ACG T275T NP_703145.1

View Full Product Details