Product Details

SNP ID
rs67312806
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:9232415 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGTTGTGCTGTACTCCCCTGGAA[C/T]CTCTGGCAACTGGACCTCCTGCAGC
Phenotype
MIM: 604874
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
A2MP1 PubMed Links

Gene Details

Gene
A2MP1
Gene Name
alpha-2-macroglobulin pseudogene 1
There are no transcripts associated with this gene.

Gene
KLRG1
Gene Name
killer cell lectin like receptor G1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005810.3 Intron NP_005801.3
XM_017018682.1 Intron XP_016874171.1
XM_017018683.1 Intron XP_016874172.1
XM_017018684.1 Intron XP_016874173.1
XM_017018685.1 Intron XP_016874174.1
Gene
LINC00987
Gene Name
long intergenic non-protein coding RNA 987
There are no transcripts associated with this gene.

Gene
MIR1244-3
Gene Name
microRNA 1244-3
There are no transcripts associated with this gene.

View Full Product Details