Product Details

SNP ID
rs74061917
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:4489254 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCTAACTTGTACATCCTTAGGAG[C/T]AGATTAAAAAGAGATTCTGAAACAG
Phenotype
MIM: 616082
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C12orf4 PubMed Links

Gene Details

Gene
C12orf4
Gene Name
chromosome 12 open reading frame 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304811.1 2208 UTR 3 NP_001291740.1
NM_020374.3 2208 UTR 3 NP_065107.1
XM_005253716.1 2208 UTR 3 XP_005253773.1
XM_006718992.3 2208 Intron XP_006719055.1

View Full Product Details