Product Details
- SNP ID
-
rs67394720
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:5438593 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTGGGTTTGTTCAGTCAGAGATTT[G/T]TTGGAAGATTTAAGGTTCTCCAACA
- Phenotype
-
MIM: 162660
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
NTF3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75379575] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NTF3
- Gene Name
- neurotrophin 3
View Full Product Details