Product Details

SNP ID
rs73407735
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:109717795 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGGCCCCACCCCCACTAATCCTT[C/T]TCACTTATCTAATTCCTGTAAGCTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM222A PubMed Links

Gene Details

Gene
FAM222A
Gene Name
family with sequence similarity 222 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032829.2 Intron NP_116218.2
XM_006719654.2 Intron XP_006719717.1
XM_017020055.1 Intron XP_016875544.1

View Full Product Details