Product Details

SNP ID
rs73408143
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:49126771 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCAAAGCCAGTTATATACTTTCT[A/G]CATTACCTCAAAGTTAGGTGGAAGT
Phenotype
MIM: 604508
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COPS2 PubMed Links

Gene Details

Gene
COPS2
Gene Name
COP9 signalosome subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143887.1 2611 UTR 3 NP_001137359.1
NM_004236.3 2611 UTR 3 NP_004227.1

View Full Product Details