Product Details

SNP ID
rs73398286
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:49423192 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCACTGGCTAAGTTTTAATTGCTT[C/T]CAATGAGGTCAGCAAAGGTATTTAT
Phenotype
MIM: 148180
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM227B PubMed Links

Gene Details

Gene
FAM227B
Gene Name
family with sequence similarity 227 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152647.2 15 Intron NP_689860.2
XM_005254213.3 15 Intron XP_005254270.1
XM_005254214.3 15 Intron XP_005254271.1
XM_005254215.3 15 Intron XP_005254272.1
XM_005254216.3 15 Intron XP_005254273.1
XM_005254219.3 15 Intron XP_005254276.1
XM_005254220.3 15 Intron XP_005254277.1
XM_006720423.2 15 Intron XP_006720486.1
XM_006720424.2 15 Intron XP_006720487.1
XM_006720426.2 15 Intron XP_006720489.1
XM_006720427.3 15 Intron XP_006720490.1
XM_011521319.2 15 Intron XP_011519621.1
XM_011521320.1 15 Intron XP_011519622.1
XM_011521321.2 15 Intron XP_011519623.1
XM_011521322.1 15 Intron XP_011519624.1
XM_011521324.1 15 Intron XP_011519626.1
XM_011521325.2 15 Intron XP_011519627.1
XM_011521326.1 15 Intron XP_011519628.1
XM_017021990.1 15 Intron XP_016877479.1
XM_017021991.1 15 Intron XP_016877480.1
XM_017021992.1 15 Intron XP_016877481.1
XM_017021993.1 15 Intron XP_016877482.1
XM_017021994.1 15 Intron XP_016877483.1
XM_017021995.1 15 Intron XP_016877484.1
XM_017021996.1 15 Intron XP_016877485.1
Gene
FGF7
Gene Name
fibroblast growth factor 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002009.3 15 UTR 5 NP_002000.1

View Full Product Details