Product Details

SNP ID
rs74015392
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:53516554 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCAATTATATAGTTCTCTTTTCT[A/C]ATTTCCCCCTCTCCTGTCCACTGCA
Phenotype
MIM: 613214
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
WDR72 PubMed Links

Gene Details

Gene
WDR72
Gene Name
WD repeat domain 72
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277176.1 4693 UTR 3 NP_001264105.1
NM_182758.3 4693 UTR 3 NP_877435.3
XM_011521433.2 4693 UTR 3 XP_011519735.1
XM_011521435.2 4693 UTR 3 XP_011519737.1
XM_011521436.2 4693 Intron XP_011519738.1
XM_011521437.2 4693 Intron XP_011519739.1
XM_017022061.1 4693 UTR 3 XP_016877550.1

View Full Product Details