Product Details

SNP ID
rs72747088
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:63199148 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCACTGTCTGTTAGCACCTCACC[C/T]ATACCAAGATTTAGAGAGAGGGGAT
Phenotype
MIM: 613532
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RAB8B PubMed Links

Gene Details

Gene
RAB8B
Gene Name
RAB8B, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016530.2 Intron NP_057614.1
XM_011521669.2 Intron XP_011519971.1
XM_017022312.1 Intron XP_016877801.1

View Full Product Details